Learn More
TARGETMOL CHEMICALS INC L-2-Hydroxyglutaric acid disod
SDP
Catalog No. 502875845
Encompass_Preferred
Also available in 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. L-2-Hydroxyglutaric acid disodium ((S)-2-Hydroxyglutaric acid disodium) is an epigenetic modifier and putative oncometabolite in renal cancer. L-2-Hydroxyglutaric acid inhibits mitochondrial creatine kinase (Mi-CK) activity(Km and Ki of 2.52 mM and 11.13 mM, respectively). Purity ≥98%
Catalog No. 50-287-5845 Supplier TARGETMOL CHEMICALS INC Supplier No. T1374850MG
Also available in 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. L-2-Hydroxyglutaric acid disodium ((S)-2-Hydroxyglutaric acid disodium) is an epigenetic modifier and putative oncometabolite in renal cancer. L-2-Hydroxyglutaric acid inhibits mitochondrial creatine kinase (Mi-CK) activity(Km and Ki of 2.52 mM and 11.13 mM, respectively). Purity ≥98%