Learn More
Medchemexpress LLC L-2-hydroxyglutaric acid (disodium) | 63512-50-5 | 99.1% | 192.08 g/mol | C5H6Na2O5 | 50 MG
SDP
Catalog No. 5000303316
Encompass_Preferred
L-2-hydroxyglutaric acid (disodium) is the disodium salt of L-2-hydroxyglutaric acid supplied for research use. It acts as an epigenetic modifier and putative oncometabolite, with reported biochemical activity against histone demethylases and mitochondrial creatine kinase. Available as a solid powder or as a 10 mM aqueous solution for laboratory use.
  • Epigenetic modifier that can inhibit histone demethylases.
  • Reported to inhibit mitochondrial creatine kinase (Mi-CK) (Km 2.52 mM; Ki 11.13 mM).
  • High purity suitable for biochemical and cellular studies.
  • Available as solid and as a 10 mM solution for convenient use.
  • Provided for research use only.
Catalog No. 50-003-03316 Supplier Medchemexpress LLC Supplier No. HYW01511450MG
L-2-hydroxyglutaric acid (disodium) is the disodium salt of L-2-hydroxyglutaric acid supplied for research use. It acts as an epigenetic modifier and putative oncometabolite, with reported biochemical activity against histone demethylases and mitochondrial creatine kinase. Available as a solid powder or as a 10 mM aqueous solution for laboratory use.
  • Epigenetic modifier that can inhibit histone demethylases.
  • Reported to inhibit mitochondrial creatine kinase (Mi-CK) (Km 2.52 mM; Ki 11.13 mM).
  • High purity suitable for biochemical and cellular studies.
  • Available as solid and as a 10 mM solution for convenient use.
  • Provided for research use only.