Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-2-Phenylglycine 98.0+%, TCI America™
SDP

Catalog No. P128825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P128825G 25 g
P1288100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P128825G Supplier TCI America Supplier No. P128825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2935-35-5
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00064403
InChI Key ZGUNAGUHMKGQNY-ZETCQYMHSA-N
Synonym h-phg-oh, l-phenylglycine, s-2-amino-2-phenylacetic acid, l-+-alpha-phenylglycine, l-2-phenylglycine, l-+-2-phenylglycine, s-phenylglycine, phenylglycine, 2s-2-amino-2-phenylacetic acid, 2s-amino phenyl acetic acid
PubChem CID 99291
ChEBI CHEBI:439819
IUPAC Name (2S)-2-amino-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)N

Specifications

Color White
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-2-Phenylglycine
EINECSNumber (3)-1441
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.