Learn More
Cayman Chemical L-3-n-ButylphthalIde 10mg
SDP
Catalog No. 502896032
Encompass_Preferred
A phthalide with antiplatelet and neuroprotective activities; inhibits ADP-, collagen-, and arachidonic acid-induced platelet aggregation ex vivo at 3-100 mg/kg; inhibits thrombus formation in a collagen and epinephrine-induced rat model of experimental thrombosis; reduces amyloid-β-induced decreases in GPX activity and decreases MDA levels in the cortex and hippocampus and improves spatial learning and memory in the Morris water maze in a rat model of Alzheimer’s disease induced by i.c.v. infusion of amyloid-β (1-40)
Catalog No. 50-289-6032 Supplier Cayman Chemical Supplier No. 900375910 MG
A phthalide with antiplatelet and neuroprotective activities; inhibits ADP-, collagen-, and arachidonic acid-induced platelet aggregation ex vivo at 3-100 mg/kg; inhibits thrombus formation in a collagen and epinephrine-induced rat model of experimental thrombosis; reduces amyloid-β-induced decreases in GPX activity and decreases MDA levels in the cortex and hippocampus and improves spatial learning and memory in the Morris water maze in a rat model of Alzheimer’s disease induced by i.c.v. infusion of amyloid-β (1-40)