Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich L-3-thienylalanine

Catalog No. CDS005041 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS005041 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS005041 Supplier Sigma Aldrich Supplier No. CDS005041100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,4-(methylenedioxy)toluene can be used as a cation scavenger for the selective cleavage of 4-(3,4-dimethoxyphenyl)benzyl (dmpbn) ethers affording the corresponding deprotected products in the presence of trifluoroacetic acid (tfa) in anhydrous ch2cl2.it can also be used as a starting material to prepare:
  • 1,3-benzodioxole-5-carboxaldehyde via photooxidation using cbr4
  • 6-methyl-1,3-benzodioxole-5-carboxaldehyde by treating with dichloromethylmethyl ether and ticl4

Specifications

Molecular Formula C7H9NO2S
Linear Formula C7H9NO2S
MDL Number MFCD00038645
Quantity 25 mL
Molecular Weight (g/mol) 171.22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.