Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [L-3-trans-(propylcarbamoyl)oxirane-2-carbonyl]-L-isoleucyl-L-proline | 134448-10-5 | MFCD00797531 | ≥95.0% | 383.44 g·mol⁻¹ | C18H29N3O6 | 1 ML
SDP

Catalog No. 5000244978
Encompass_Preferred

CA-074 is a potent, selective inhibitor of cathepsin B (Ki = 2-5 nM) commonly supplied as a 10 mM solution in DMSO for use in biochemical and cellular research. The compound has molecular formula C18H29N3O6 and a molecular weight of 383.44 g·mol⁻¹.

  • Potent cathepsin B inhibition with Ki = 2-5 nM.
  • Supplied as a 10 mM solution, 1 mL vial, ready for in vitro use.
  • Soluble in DMSO; example in vitro stock concentrations provided by the manufacturer.
  • Storage guidance: powder -20 °C; in solvent -80 °C (up to 6 months) or -20 °C (up to 1 month).
  • Molecular formula C18H29N3O6; molecular weight 383.44 g·mol⁻¹.
  • Intended for research use only (not for human or veterinary use).

Catalog No. 50-002-44978 Supplier Medchemexpress LLC Supplier No. HY10335010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CA-074 is a potent, selective inhibitor of cathepsin B (Ki = 2-5 nM) commonly supplied as a 10 mM solution in DMSO for use in biochemical and cellular research. The compound has molecular formula C18H29N3O6 and a molecular weight of 383.44 g·mol⁻¹.

  • Potent cathepsin B inhibition with Ki = 2-5 nM.
  • Supplied as a 10 mM solution, 1 mL vial, ready for in vitro use.
  • Soluble in DMSO; example in vitro stock concentrations provided by the manufacturer.
  • Storage guidance: powder -20 °C; in solvent -80 °C (up to 6 months) or -20 °C (up to 1 month).
  • Molecular formula C18H29N3O6; molecular weight 383.44 g·mol⁻¹.
  • Intended for research use only (not for human or veterinary use).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.