Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-364,373 | 103342-82-1 | 99.7% | 397.44 g·mol⁻¹ | C25H20FN3O | 5 MG
SDP

Catalog No. 5000263725
Encompass_Preferred

L-364,373 (R-L3) is a small-molecule activator of voltage-gated Kv7.1 (KCNQ1)/mink potassium channels. It enhances the slow delayed rectifier potassium current (Iks), shortens action potential duration in cardiac myocytes, and suppresses early afterdepolarizations, making it useful for electrophysiology and cardiovascular research.

  • CAS number: 103342-82-1.
  • Purity: 99.67%.
  • Molecular weight: 397.44 g·mol⁻¹.
  • Chemical formula: C25H20FN3O.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Solubility: DMSO 100 mg/mL; in vivo formulations documented for dosing.
  • Pack size: 5 mg available.

Catalog No. 50-002-63725 Supplier Medchemexpress LLC Supplier No. HY1085915MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-364,373 (R-L3) is a small-molecule activator of voltage-gated Kv7.1 (KCNQ1)/mink potassium channels. It enhances the slow delayed rectifier potassium current (Iks), shortens action potential duration in cardiac myocytes, and suppresses early afterdepolarizations, making it useful for electrophysiology and cardiovascular research.

  • CAS number: 103342-82-1.
  • Purity: 99.67%.
  • Molecular weight: 397.44 g·mol⁻¹.
  • Chemical formula: C25H20FN3O.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Solubility: DMSO 100 mg/mL; in vivo formulations documented for dosing.
  • Pack size: 5 mg available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.