Learn More
Medchemexpress LLC HY-108677 10mg Medchemexpress, L-368,899 hydrochloride CAS:160312-62-9 Purity:>98%
SDP
Catalog No. 501968605
Encompass_Preferred
Medchemexpress, HY-108677 10mg L-368,899 HCl CAS:160312-62-9 L-368,899 hydrochloride is a potent, selective, orally bioavailable, non-peptide oxytocin receptor antagonist, with IC50s of 8.9 nM and 26 nM for rat uterus and human uterus oxytocin receptor, respectively, used as a tocolytic agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8605 Supplier Medchemexpress LLC Supplier No. HY10867710MG
Medchemexpress, HY-108677 10mg L-368,899 HCl CAS:160312-62-9 L-368,899 hydrochloride is a potent, selective, orally bioavailable, non-peptide oxytocin receptor antagonist, with IC50s of 8.9 nM and 26 nM for rat uterus and human uterus oxytocin receptor, respectively, used as a tocolytic agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.