Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-368,899 hydrochloride | 160312-62-9 | 100.0% | 591.23 | 10 MG
SDP

Catalog No. 5000344708
Encompass_Preferred

L-368,899 hydrochloride is a potent, selective, orally bioavailable, non-peptide oxytocin receptor antagonist. It has IC50s of 8.9 nM for rat uterus oxytocin receptor and 26 nM for human uterus oxytocin receptor. It is used as a tocolytic agent.

  • Potent oxytocin receptor antagonist
  • Selective action
  • Orally bioavailable
  • Non-peptide structure
  • IC50s of 8.9 nM for rat uterus oxytocin receptor and 26 nM for human uterus oxytocin receptor
  • Used as a tocolytic agent
  • Soluble in DMSO (100 mg/mL)
  • Soluble in H2O (5 mg/mL)
  • Storage at 4°C sealed, away from moisture
  • In solvent storage at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-003-44708 Supplier Medchemexpress LLC Supplier No. HY1086771MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-368,899 hydrochloride is a potent, selective, orally bioavailable, non-peptide oxytocin receptor antagonist. It has IC50s of 8.9 nM for rat uterus oxytocin receptor and 26 nM for human uterus oxytocin receptor. It is used as a tocolytic agent.

  • Potent oxytocin receptor antagonist
  • Selective action
  • Orally bioavailable
  • Non-peptide structure
  • IC50s of 8.9 nM for rat uterus oxytocin receptor and 26 nM for human uterus oxytocin receptor
  • Used as a tocolytic agent
  • Soluble in DMSO (100 mg/mL)
  • Soluble in H2O (5 mg/mL)
  • Storage at 4°C sealed, away from moisture
  • In solvent storage at -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.