Learn More
Medchemexpress LLC L-368,899 hydrochloride | 160312-62-9 | 99.9% | 591.23 | 25 MG
SDP
Catalog No. 5000347412
Encompass_Preferred
L-368,899 hydrochloride is a potent, selective, orally bioavailable, non-peptide oxytocin receptor antagonist. It is used as a tocolytic agent and shows less activity on VP receptors in human and rat liver and kidney.
  • Potent, orally bioavailable, non-peptide oxytocin receptor antagonist
  • IC50s of 8.9 nM for rat uterus oxytocin receptor
  • IC50s of 26 nM for human uterus oxytocin receptor
  • Used as a tocolytic agent
  • Less active on VP receptor in human and rat liver and kidney
  • Exhibits similar pharmacokinetics in rats and dogs
  • Orally available with demonstrated bioavailability in rats
Catalog No. 50-003-47412 Supplier Medchemexpress LLC Supplier No. 25MGHY108677
L-368,899 hydrochloride is a potent, selective, orally bioavailable, non-peptide oxytocin receptor antagonist. It is used as a tocolytic agent and shows less activity on VP receptors in human and rat liver and kidney.
  • Potent, orally bioavailable, non-peptide oxytocin receptor antagonist
  • IC50s of 8.9 nM for rat uterus oxytocin receptor
  • IC50s of 26 nM for human uterus oxytocin receptor
  • Used as a tocolytic agent
  • Less active on VP receptor in human and rat liver and kidney
  • Exhibits similar pharmacokinetics in rats and dogs
  • Orally available with demonstrated bioavailability in rats