Learn More
Medchemexpress LLC L-368,899 hydrochloride | 160312-62-9 | 99.97% | 591.23 | 100 MG
SDP
Catalog No. 5000344524
Encompass_Preferred
L-368,899 hydrochloride is a potent, selective, orally bioavailable, non-peptide oxytocin receptor antagonist. It has IC50s of 8.9 nM for rat uterus and 26 nM for human uterus oxytocin receptor. This compound is used as a tocolytic agent.
  • Potent oxytocin receptor antagonist.
  • Selective for oxytocin receptor.
  • Orally bioavailable.
  • Non-peptide structure.
  • Used as a tocolytic agent.
  • Less active on VP receptor in human liver and kidney, and rat liver and kidney.
  • Exhibits similar pharmacokinetics in rats and dogs with a t1/2 of 2 hours after a single IV injection.
  • Plasma clearance between 23 and 36 ml/min/kg in rats or dogs.
  • Vdss values of 2.0 and 2.6 liters/kg for rats and 3.4 to 4.9 liters/kg for dogs.
  • Oral bioavailabilities in rats: 14% (female) and 18% (male) at 5 mg/kg; 17% (female) and 41% (male) at 25 mg/kg.
Catalog No. 50-003-44524 Supplier Medchemexpress LLC Supplier No. HY10867710MM/1ML
L-368,899 hydrochloride is a potent, selective, orally bioavailable, non-peptide oxytocin receptor antagonist. It has IC50s of 8.9 nM for rat uterus and 26 nM for human uterus oxytocin receptor. This compound is used as a tocolytic agent.
  • Potent oxytocin receptor antagonist.
  • Selective for oxytocin receptor.
  • Orally bioavailable.
  • Non-peptide structure.
  • Used as a tocolytic agent.
  • Less active on VP receptor in human liver and kidney, and rat liver and kidney.
  • Exhibits similar pharmacokinetics in rats and dogs with a t1/2 of 2 hours after a single IV injection.
  • Plasma clearance between 23 and 36 ml/min/kg in rats or dogs.
  • Vdss values of 2.0 and 2.6 liters/kg for rats and 3.4 to 4.9 liters/kg for dogs.
  • Oral bioavailabilities in rats: 14% (female) and 18% (male) at 5 mg/kg; 17% (female) and 41% (male) at 25 mg/kg.