Learn More
TARGETMOL CHEMICALS INC L-368 899 hydrochloride 2MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T117942MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. L-368,899 hydrochloride is an orally available and selective nonpeptide oxytocin receptor (OXTR) antagonist for the prevention of preterm labor.L-368,899 hydrochloride prevents oxytocin-induced anti-anomalous pain and anti-nausea, and may be useful for the study of depression. Purity 99.59%
Catalog No. 50-287-9827