Learn More
Medchemexpress LLC L-371,257 | 162042-44-6 | 99.3% | 507.58 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1501050MG
Encompass_Preferred
L-371,257 is an orally bioavailable, non-blood-brain barrier penetrant, selective, and competitive antagonist of the oxytocin receptor. It exhibits high affinity for both the oxytocin receptor (Ki=19 nM) and the vasopressin V1a receptor (Ki=3.7 nM). This compound has been shown to stimulate weight gain in rats when administered systemically at doses of 0.5 and 1.0 mg/kg via intraperitoneal injection.
  • Orally bioavailable.
  • Does not penetrate the blood-brain barrier.
  • Selective and competitive antagonist of the oxytocin receptor.
  • High affinity at both the oxytocin receptor (Ki=19 nM) and vasopressin V1a receptor (Ki=3.7 nM).
  • Stimulates weight gain in rats.
Catalog No. 50-003-60188