Learn More
Medchemexpress LLC L-371,257 | 162042-44-6 | 99.34 % | 507.58 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15010100MG
Encompass_Preferred
L-371,257 is an orally bioavailable, non-blood-brain barrier penetrant compound that functions as a selective and competitive antagonist of the oxytocin receptor. It exhibits high affinity for both the oxytocin receptor (Ki=19 nM) and the vasopressin V1a receptor (Ki=3.7 nM).
  • Orally bioavailable
  • Non-blood-brain barrier penetrant
  • Selective and competitive oxytocin receptor antagonist (pA2=8.4)
  • High affinity for oxytocin receptor (Ki=19 nM)
  • High affinity for vasopressin V1a receptor (Ki=3.7 nM)
  • Stimulates weight gain in rat models
  • Exhibits activity at human oxytocin receptor in CHO cells (EC50 = 4.6 nM)
Catalog No. 50-003-61087