Learn More
Medchemexpress LLC L-371,257 | 162042-44-6 | 99.34% | 507.58 | 25 MG
SDP
Catalog No. 5000360767
Encompass_Preferred
L-371,257 is an orally bioavailable, non-blood-brain barrier penetrant, selective and competitive antagonist of the oxytocin receptor. It exhibits high affinity at both the oxytocin receptor (Ki=19 nM) and vasopressin V1a receptor (Ki=3.7 nM). It is for research use only.
  • Selective and competitive antagonist of oxytocin receptor
  • High affinity for oxytocin receptor (Ki=19 nM)
  • High affinity for vasopressin V1a receptor (Ki=3.7 nM)
  • Orally bioavailable
  • Non-blood-brain barrier penetrant
  • Stimulates weight gain in rats
  • White to off-white solid appearance
Catalog No. 50-003-60767 Supplier Medchemexpress LLC Supplier No. 25MGHY15010
L-371,257 is an orally bioavailable, non-blood-brain barrier penetrant, selective and competitive antagonist of the oxytocin receptor. It exhibits high affinity at both the oxytocin receptor (Ki=19 nM) and vasopressin V1a receptor (Ki=3.7 nM). It is for research use only.
  • Selective and competitive antagonist of oxytocin receptor
  • High affinity for oxytocin receptor (Ki=19 nM)
  • High affinity for vasopressin V1a receptor (Ki=3.7 nM)
  • Orally bioavailable
  • Non-blood-brain barrier penetrant
  • Stimulates weight gain in rats
  • White to off-white solid appearance