Learn More
Medchemexpress LLC HY-15010 5mg Medchemexpress, L-371,257 CAS:162042-44-6 Purity:>98%
SDP
Catalog No. 501932720
Encompass_Preferred
Medchemexpress, HY-15010 5mg L-371,257 CAS:162042-44-6 L-371,257 is an orally bioavailable, non-blood-brain barrier penetrant, selective and competitive antagonist of oxytocin receptor (pA2=8.4) with high affinity at both the oxytocin receptor ( K i =19 nM) and vasopressin V1a receptor ( K i =3.7 nM) [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2720 Supplier Medchemexpress LLC Supplier No. HY150105MG
Medchemexpress, HY-15010 5mg L-371,257 CAS:162042-44-6 L-371,257 is an orally bioavailable, non-blood-brain barrier penetrant, selective and competitive antagonist of oxytocin receptor (pA2=8.4) with high affinity at both the oxytocin receptor ( K i =19 nM) and vasopressin V1a receptor ( K i =3.7 nM) [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.