Learn More
Cayman Chemical l-3718-37humntrIflu 10mg
SDP
Catalog No. 502914961
Encompass_Preferred
An antimicrobial peptide fragment of LL-37; active against S. aureus and C. albicans with minimum lethal concentrations of 4 and 10 µM, respectively; inhibits the growth of E. histolytica trophozoites from 10-50 µM
Catalog No. 50-291-4961 Supplier Cayman Chemical Supplier No. 3695010 MG
An antimicrobial peptide fragment of LL-37; active against S. aureus and C. albicans with minimum lethal concentrations of 4 and 10 µM, respectively; inhibits the growth of E. histolytica trophozoites from 10-50 µM