Learn More
Medchemexpress LLC L-372662 | 162045-26-3 | C₃₃H₃₈N₄O₆ | 5 MG
SDP
Catalog No. 5000374878
Encompass_Preferred
L-372662 is a potent and orally active non-peptide oxytocin antagonist with a Ki value of 4.8. It demonstrates selectivity to OTR:V1aR and acts as an antagonist of oxytocin-induced uterine contractions. This compound exhibits good oral bioavailability and aqueous solubility.
  • Potent and orally active non-peptide oxytocin antagonist
  • Ki value of 4.8
  • Kd value for wild-type hOTR is 5.8 nM
  • Kd value for [A318G]OTR is 73 nM
  • Selectivity to OTR:V1aR
  • Antagonist of oxytocin-induced uterine contractions
  • Oral bioavailability (F = 90% in dogs)
  • Aqueous solubility (10 mg/mL)
  • For research use only
Catalog No. 50-003-74878 Supplier Medchemexpress LLC Supplier No. HY150115MG
L-372662 is a potent and orally active non-peptide oxytocin antagonist with a Ki value of 4.8. It demonstrates selectivity to OTR:V1aR and acts as an antagonist of oxytocin-induced uterine contractions. This compound exhibits good oral bioavailability and aqueous solubility.
  • Potent and orally active non-peptide oxytocin antagonist
  • Ki value of 4.8
  • Kd value for wild-type hOTR is 5.8 nM
  • Kd value for [A318G]OTR is 73 nM
  • Selectivity to OTR:V1aR
  • Antagonist of oxytocin-induced uterine contractions
  • Oral bioavailability (F = 90% in dogs)
  • Aqueous solubility (10 mg/mL)
  • For research use only