Learn More
Medchemexpress LLC L-4-Chlorophenylglycine | 67336-19-0 | 185.61 | 100 G
SDP
Catalog No. 5000449279
Encompass_Preferred
L-4-Chlorophenylglycine is a derivative of Glycine intended for research use only. It is utilized as an ergogenic supplement, influencing anabolic hormone secretion, supplying fuel during exercise, enhancing mental performance, and helping prevent muscle damage. It appears as a solid, white to off-white in color.
  • Derivative of Glycine
  • For research use only
  • Used as an ergogenic supplement
  • Influences anabolic hormone secretion
  • Supplies fuel during exercise
  • Enhances mental performance during stress
  • Helps prevent exercise-induced muscle damage
  • Solid, white to off-white appearance
Catalog No. 50-004-49279 Supplier Medchemexpress LLC Supplier No. HYW005720100G
L-4-Chlorophenylglycine is a derivative of Glycine intended for research use only. It is utilized as an ergogenic supplement, influencing anabolic hormone secretion, supplying fuel during exercise, enhancing mental performance, and helping prevent muscle damage. It appears as a solid, white to off-white in color.
  • Derivative of Glycine
  • For research use only
  • Used as an ergogenic supplement
  • Influences anabolic hormone secretion
  • Supplies fuel during exercise
  • Enhances mental performance during stress
  • Helps prevent exercise-induced muscle damage
  • Solid, white to off-white appearance