Learn More
Selleck Chemical LLC L-5-Methyltetrahydrofolate calcium S5264-1g
SDP
Catalog No. 503111926
Encompass_Preferred
Calcium L-5-methyltetrahydrofolate (L-5-MTHF-Ca) is a synthetic derivative of folic acid the predominant naturally occurring form of folate
Catalog No. 50-311-1926 Supplier Selleck Chemical LLC Supplier No. S52641G
Calcium L-5-methyltetrahydrofolate (L-5-MTHF-Ca) is a synthetic derivative of folic acid the predominant naturally occurring form of folate