Learn More
Selleck Chemical LLC L-685 458
SDP
Catalog No. 5000000258
Encompass_Preferred
L-685 458 is a specific and potent inhibitor of A beta PP gamma-secretase activity with Ki of 17 nM
Catalog No. 50-000-00258 Supplier Selleck Chemical LLC Supplier No. S76735MG
L-685 458 is a specific and potent inhibitor of A beta PP gamma-secretase activity with Ki of 17 nM