Learn More
Apexbio Technology LLC L-690,330 142523-38-4 10mg
SDP
Catalog No. 502240123
Encompass_Preferred
L-690 330 (CAS 142523-38-4) is a competitive inhibitor of inositol monophosphatase (IMPase) exhibiting a Ki between 0 2 and 2 M depending on the enzyme source IMPase catalyzes the hydrolysis of inositol monophosphates yielding free inositol for phosphatidylinositol (PI) cycle signaling In M1 CHO cells L-690 330 did not elevate [3H]InsP1 levels alone but attenuated carbachol-stimulated increases producing effects up to 40% of those observed with lithium In vivo L-690 330 administration led to significant Ins(1)P accumulation in mouse brain following muscarinic agonist stimulation These properties make L-690 330 a valuable tool for probing inositol signaling pathways and studying IMPase-related neurological processes
This item is not returnable. View return policy
Catalog No. 50-224-0123 Supplier Apexbio Technology LLC Supplier No. B501110
L-690 330 (CAS 142523-38-4) is a competitive inhibitor of inositol monophosphatase (IMPase) exhibiting a Ki between 0 2 and 2 M depending on the enzyme source IMPase catalyzes the hydrolysis of inositol monophosphates yielding free inositol for phosphatidylinositol (PI) cycle signaling In M1 CHO cells L-690 330 did not elevate [3H]InsP1 levels alone but attenuated carbachol-stimulated increases producing effects up to 40% of those observed with lithium In vivo L-690 330 administration led to significant Ins(1)P accumulation in mouse brain following muscarinic agonist stimulation These properties make L-690 330 a valuable tool for probing inositol signaling pathways and studying IMPase-related neurological processes