Learn More
Medchemexpress LLC L-692429 | 145455-23-8 | 99.9% | 509.60 | 50 MG
SDP
Catalog No. 5000343262
Encompass_Preferred
L-692429 (MK-0751) is a benzolactam derivative and a nonpeptidyl growth hormone secretagogue (GHS) agonist. It binds to G protein-coupled receptor with a Ki of 63 nM. This product is for research use only.
  • Stimulates intracellular calcium release
  • Activates inositol phosphate (IP) turnover
  • Increases cAMP-responsive element binding protein (CREB) activity
  • Enhances serum-responsive element activity
  • Promotes bioluminescence resonance energy transfer (BRET) activity
  • Suitable for laboratory centrifugation and storage
Catalog No. 50-003-43262 Supplier Medchemexpress LLC Supplier No. HY1095710MG
L-692429 (MK-0751) is a benzolactam derivative and a nonpeptidyl growth hormone secretagogue (GHS) agonist. It binds to G protein-coupled receptor with a Ki of 63 nM. This product is for research use only.
  • Stimulates intracellular calcium release
  • Activates inositol phosphate (IP) turnover
  • Increases cAMP-responsive element binding protein (CREB) activity
  • Enhances serum-responsive element activity
  • Promotes bioluminescence resonance energy transfer (BRET) activity
  • Suitable for laboratory centrifugation and storage