Learn More
Apexbio Technology LLC L-693,403 maleate 207455-21-8 50mg
SDP
Catalog No. 5000091854
Encompass_Preferred
L-693 403 maleate (CAS 207455-21-8) is a small molecule exhibiting high affinity and selectivity for sigma ( ) receptors By modulating receptor-mediated signaling pathways it influences neuronal signal transduction at the molecular level Preclinical studies indicate that L-693 403 maleate possesses analgesic properties likely through its regulatory effects on neural activity Owing to these pharmacological characteristics it is utilized in exploratory research on neurological disorders supporting its value as a tool compound in the investigation of neural modulation mechanisms
Catalog No. 50-000-91854 Supplier Apexbio Technology LLC Supplier No. B500250
L-693 403 maleate (CAS 207455-21-8) is a small molecule exhibiting high affinity and selectivity for sigma ( ) receptors By modulating receptor-mediated signaling pathways it influences neuronal signal transduction at the molecular level Preclinical studies indicate that L-693 403 maleate possesses analgesic properties likely through its regulatory effects on neural activity Owing to these pharmacological characteristics it is utilized in exploratory research on neurological disorders supporting its value as a tool compound in the investigation of neural modulation mechanisms