Learn More
Bachem H-4-(7-Hydroxycoumarin-4-yl)-Abu-OH
SDP
Catalog No. 501947208
Encompass_Preferred
H-4-(7-Hydroxycoumarin-4-yl)-Abu-OH, 905442-42-4, C13H13NO5, Mr 263.25, 1g. H-(7-Hydroxycoumarin-4-yl)-ethyl-Gly-OH, H-(Umbellifer-4-yl)-ethyl-Gly-OH. L-(7-hydroxycoumarin-4-yl)ethylglycine (umbelliferylethylglycine) has been incorporated into proteins as a fluorescent tag.
Catalog No. 50-194-7208 Supplier Bachem Supplier No. 4059541.1000
Add to Cart
Add to Cart
H-4-(7-Hydroxycoumarin-4-yl)-Abu-OH, 905442-42-4, C13H13NO5, Mr 263.25, 1g. H-(7-Hydroxycoumarin-4-yl)-ethyl-Gly-OH, H-(Umbellifer-4-yl)-ethyl-Gly-OH. L-(7-hydroxycoumarin-4-yl)ethylglycine (umbelliferylethylglycine) has been incorporated into proteins as a fluorescent tag.