Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-732138 | 148451-96-1 | MFCD00237267 | 99.9% | 472.38 g/mol | C22H18F6N2O3 | 1 ML
SDP

Catalog No. 5000212860
Encompass_Preferred

L-732138 is a selective, potent, competitive neurokinin-1 (NK-1) receptor antagonist supplied as a 10 mM solution in DMSO (1 mL). It is provided at high purity for use in receptor pharmacology, signaling studies, and compound screening where a defined, ready-to-use reagent is required.

  • High purity (99.9%) suitable for analytical and biological assays.
  • Ready-to-use 10 mM solution in DMSO for convenient dosing and dilution.
  • Molecular weight 472.38 g/mol and defined chemical identity for reproducibility.
  • Storage guidance provided to maintain stability in powder and solution forms.
  • Appropriate for NK-1 receptor pharmacology and related cellular assays.

Catalog No. 50-002-12860 Supplier Medchemexpress LLC Supplier No. HY10124910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-732138 is a selective, potent, competitive neurokinin-1 (NK-1) receptor antagonist supplied as a 10 mM solution in DMSO (1 mL). It is provided at high purity for use in receptor pharmacology, signaling studies, and compound screening where a defined, ready-to-use reagent is required.

  • High purity (99.9%) suitable for analytical and biological assays.
  • Ready-to-use 10 mM solution in DMSO for convenient dosing and dilution.
  • Molecular weight 472.38 g/mol and defined chemical identity for reproducibility.
  • Storage guidance provided to maintain stability in powder and solution forms.
  • Appropriate for NK-1 receptor pharmacology and related cellular assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.