Learn More
Medchemexpress LLC HY-101249 1mg Medchemexpress, L-732138 CAS:148451-96-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1012491MG
Encompass_Preferred
Medchemexpress, HY-101249 1mg L-732138 CAS:148451-96-1 L-732138 is a selective, potent and competitive neurokinin-1 (NK-1) receptor antagonist with an IC50 of 2.3 nM. L-732138 has 200-fold more potent in cloned human NK-1 receptors than cloned rat NK-1 receptors, and has > 1000-fold more potent than human NK-2 and NK-3 receptors. L-732138 can reduce hyperalgesia and has antitumor action. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7815