Learn More
Medchemexpress LLC L-733060 hydrochloride | 148687-76-7 | 99.9% | 439.82 | 1 MG
SDP
Catalog No. 5000365247
Encompass_Preferred
L-733060 hydrochloride is a selective neurokinin-1 (NK-1) receptor antagonist. It primarily regulates pain transmission and neural plasticity by blocking the binding of Substance P to the NK-1 receptor. It blocks the promoting effect of Substance P on long-term potentiation (LTP) in the hippocampus and reverses orofacial hyperalgesia induced by experimental occlusal interference (EOI) in rats. Additionally, it inhibits neurogenic plasma extravasation and acts as an anti-tumor agent, making it suitable for studying chronic orofacial pain.
  • Selective neurokinin-1 (NK-1) receptor antagonist
  • Regulates pain transmission and neural plasticity
  • Blocks Substance P binding to NK-1 receptor
  • Reverses orofacial hyperalgesia
  • Inhibits neurogenic plasma extravasation
  • Acts as an anti-tumor agent
  • Suitable for studying chronic orofacial pain
Catalog No. 50-003-65247 Supplier Medchemexpress LLC Supplier No. 1MGHY14406A
L-733060 hydrochloride is a selective neurokinin-1 (NK-1) receptor antagonist. It primarily regulates pain transmission and neural plasticity by blocking the binding of Substance P to the NK-1 receptor. It blocks the promoting effect of Substance P on long-term potentiation (LTP) in the hippocampus and reverses orofacial hyperalgesia induced by experimental occlusal interference (EOI) in rats. Additionally, it inhibits neurogenic plasma extravasation and acts as an anti-tumor agent, making it suitable for studying chronic orofacial pain.
  • Selective neurokinin-1 (NK-1) receptor antagonist
  • Regulates pain transmission and neural plasticity
  • Blocks Substance P binding to NK-1 receptor
  • Reverses orofacial hyperalgesia
  • Inhibits neurogenic plasma extravasation
  • Acts as an anti-tumor agent
  • Suitable for studying chronic orofacial pain