Learn More
Medchemexpress LLC L-741626 | 81226-60-0 | 99.0% | 340.85 | C20H21ClN2O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10134810MM 1ML
Encompass_Preferred
L-741626 is a selective D2 dopamine receptor antagonist supplied as a 10 mM solution in DMSO (1 mL) for research use. It exhibits Ki values of 2.4 nM for human D2, 100 nM for D3, and 220 nM for D4. The compound has CAS 81226-60-0 and a reported purity of 99.0%. Store powder at -20°C; in solution store at -80°C for up to 6 months or at -20°C for short-term storage.
  • Selective D2 receptor antagonism with low nanomolar potency.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • High chemical purity (99.0%) for reliable results.
  • Compact 1 mL volume suitable for small-scale assays.
  • Clear storage guidelines to preserve stability.
  • Suitable for receptor pharmacology and neuroscience studies.
Catalog No. 50-002-13149