Learn More
Cayman Chemical L-742 001hydrochlorIde 10mg
SDP
Catalog No. 502942556
Encompass_Preferred
An inhibitor of influenza PA endonuclease (IC50 = 0.5 µM); reduces viral titers in PR8 influenza-infected MDCK cells at 10 µM
Catalog No. 50-294-2556 Supplier Cayman Chemical Supplier No. 4092010 MG
An inhibitor of influenza PA endonuclease (IC50 = 0.5 µM); reduces viral titers in PR8 influenza-infected MDCK cells at 10 µM