Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-742001 (hydrochloride) | 174605-64-2 | 99.0% | 450.35 | C23H25Cl2NO4 | 1 MG
SDP

Catalog No. 5000308294
Encompass_Preferred

L-742001 hydrochloride is a small-molecule inhibitor of the influenza virus PA endonuclease, supplied as a high-purity research reagent for in vitro virology and mechanism-of-action studies.

  • High purity: 99.0% (analytical).
  • Molecular weight: 450.35 g·mol⁻¹.
  • CAS number: 174605-64-2.
  • Mechanism: influenza PA endonuclease inhibitor.
  • Reported activity: EC90 = 4.3 μM for vRNP activity in HEK293T cells.
  • Suitable for in vitro virology and antiviral mechanism studies.
  • Available in small research pack sizes, including 1 mg.

Catalog No. 50-003-08294 Supplier Medchemexpress LLC Supplier No. HY1183681MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-742001 hydrochloride is a small-molecule inhibitor of the influenza virus PA endonuclease, supplied as a high-purity research reagent for in vitro virology and mechanism-of-action studies.

  • High purity: 99.0% (analytical).
  • Molecular weight: 450.35 g·mol⁻¹.
  • CAS number: 174605-64-2.
  • Mechanism: influenza PA endonuclease inhibitor.
  • Reported activity: EC90 = 4.3 μM for vRNP activity in HEK293T cells.
  • Suitable for in vitro virology and antiviral mechanism studies.
  • Available in small research pack sizes, including 1 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.