Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-742001 hydrochloride | 174605-64-2 | 99.0% | 450.35 | C23H25Cl2NO4 | 5 MG
SDP

Catalog No. 5000227464
Encompass_Preferred

L-742001 hydrochloride is a research-grade small-molecule inhibitor of influenza virus PA endonuclease, supplied as a high-purity solid for in vitro studies. It reduces vRNP activity in HEK293T cells with an EC90 of 4.3 μM and is intended for laboratory research use only.

  • Inhibits influenza PA endonuclease, EC90 of 4.3 μM for vRNP activity in HEK293T cells.
  • High purity (99.0%) solid suitable for research applications.
  • Molecular weight 450.35 and formula C23H25Cl2NO4.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-27464 Supplier Medchemexpress LLC Supplier No. HY1183685MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-742001 hydrochloride is a research-grade small-molecule inhibitor of influenza virus PA endonuclease, supplied as a high-purity solid for in vitro studies. It reduces vRNP activity in HEK293T cells with an EC90 of 4.3 μM and is intended for laboratory research use only.

  • Inhibits influenza PA endonuclease, EC90 of 4.3 μM for vRNP activity in HEK293T cells.
  • High purity (99.0%) solid suitable for research applications.
  • Molecular weight 450.35 and formula C23H25Cl2NO4.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.