Learn More
Enzo Life Sciences L-744,832 (5mg). CAS: 1177806-11-9
SDP
Supplier:  Enzo Life Sciences BMLG2420005
Encompass_Preferred
Ras farnesyltransferase inhibitor. Induces tumor regression in transgenic mice with multiple oncogenic mutations by mediating alterations in both cell cycle control and apoptosis. Induces p21 expression and arrests cells at G1. Causes enhanced mitotic sensitivity to taxol. Purity: ≥98% (TLC). Solubility: Soluble in DMSO, water, or acetonitrile. Long Term Storage: -20°C.
Catalog No. 50-201-0872