Learn More
Medchemexpress LLC L-745870 hydrochloride | 1173023-36-3 | 99.9% | C18H20Cl2N4 | 10MG
SDP
Catalog No. 5000261398
Encompass_Preferred
L-745870 hydrochloride is the hydrochloride salt of a selective dopamine D4 receptor antagonist used in pharmacology research. It is brain-penetrant, orally active, and is employed to investigate D4 receptor activity and related neuropharmacological effects.
  • Potent and selective dopamine D4 receptor antagonist (Ki 0.43 nM).
  • Brain-penetrant and orally active for in vivo studies.
  • High chemical purity suitable for research use (99.9%).
  • Available as a solid or as a DMSO solution for flexibility in assays.
  • Offered in multiple small-scale package sizes for preclinical experiments.
Catalog No. 50-002-61398 Supplier Medchemexpress LLC Supplier No. HY14325B10MG
L-745870 hydrochloride is the hydrochloride salt of a selective dopamine D4 receptor antagonist used in pharmacology research. It is brain-penetrant, orally active, and is employed to investigate D4 receptor activity and related neuropharmacological effects.
  • Potent and selective dopamine D4 receptor antagonist (Ki 0.43 nM).
  • Brain-penetrant and orally active for in vivo studies.
  • High chemical purity suitable for research use (99.9%).
  • Available as a solid or as a DMSO solution for flexibility in assays.
  • Offered in multiple small-scale package sizes for preclinical experiments.