Learn More
Medchemexpress LLC L-745870 hydrochloride | 1173023-36-3 | 99.91% | C18H20Cl2N4 | 1 ML
SDP
Catalog No. 5000354237
Encompass_Preferred
L-745870 hydrochloride is a potent, selective, brain-penetrant, and orally active dopamine D4 receptor antagonist. It exhibits high affinity for D4 receptors, with weaker affinity for D2 and D3 receptors, and moderate affinity for 5-HT2 receptors, sigma sites, and α-adrenoceptors. This compound demonstrates good pharmacokinetic properties and excellent brain penetration in vivo.
  • Potent, selective, brain-penetrant, and orally active dopamine D4 receptor antagonist
  • High affinity for D4 receptors (Ki of 0.43 nM)
  • Weaker affinity for D2 and D3 receptors
  • Moderate affinity for 5-HT2 receptors, sigma sites, and α-adrenoceptors
  • Good pharmacokinetic properties with 20-60% oral bioavailability
  • Plasma t1/2 of 2.1-2.8 hours in rats and monkeys
  • Excellent brain penetration in rats
Catalog No. 50-003-54237 Supplier Medchemexpress LLC Supplier No. HY14325B10MM/1ML
L-745870 hydrochloride is a potent, selective, brain-penetrant, and orally active dopamine D4 receptor antagonist. It exhibits high affinity for D4 receptors, with weaker affinity for D2 and D3 receptors, and moderate affinity for 5-HT2 receptors, sigma sites, and α-adrenoceptors. This compound demonstrates good pharmacokinetic properties and excellent brain penetration in vivo.
  • Potent, selective, brain-penetrant, and orally active dopamine D4 receptor antagonist
  • High affinity for D4 receptors (Ki of 0.43 nM)
  • Weaker affinity for D2 and D3 receptors
  • Moderate affinity for 5-HT2 receptors, sigma sites, and α-adrenoceptors
  • Good pharmacokinetic properties with 20-60% oral bioavailability
  • Plasma t1/2 of 2.1-2.8 hours in rats and monkeys
  • Excellent brain penetration in rats