Learn More
Abcam L-745870 trihydrochloride, brain penetrant D4 antagonist, 50MG
Catalog No. 502707439
Encompass_Preferred
MW 436.21 g/mol, Purity >99%. Highly potent and selective D₄ antagonist (Ki values are 0.43, 2300 and 960 nM at D₄, D₃ and D₂, respectively). Brain permeable. Inhibits cell death and shows neuroprotective effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7439 Supplier Abcam Supplier No. AB12059850MG
MW 436.21 g/mol, Purity >99%. Highly potent and selective D₄ antagonist (Ki values are 0.43, 2300 and 960 nM at D₄, D₃ and D₂, respectively). Brain permeable. Inhibits cell death and shows neuroprotective effects.
The product is subject to the following: Abcam Restricted Use Statement