Learn More
TARGETMOL CHEMICALS INC L 756423 5MG
SDP
Catalog No. 502580813
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. L 756423 (L756423) is a novel potent and selective and competitive HIV protease inhibitor with a Ki value of 0.049 nM for HIV-1 PR.L 756423 blocks the spread of HIV(IIIb)-infected MT25 lymphocytes at 0.50-0.1 nM and can be used for the study of Acquired Immunodeficiency Syndrome (AIDS). purity: 100%
Catalog No. 50-258-0813 Supplier TARGETMOL CHEMICALS INC Supplier No. T24362
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. L 756423 (L756423) is a novel potent and selective and competitive HIV protease inhibitor with a Ki value of 0.049 nM for HIV-1 PR.L 756423 blocks the spread of HIV(IIIb)-infected MT25 lymphocytes at 0.50-0.1 nM and can be used for the study of Acquired Immunodeficiency Syndrome (AIDS). purity: 100%