Learn More
Cayman Chemical L-778,123 (hydrochloride), Purity: ≥98%, CAS: 253863-00-2, 25 mg
SDP
Catalog No. 502994563
Encompass_Preferred
A dual inhibitor of FTase and GGTase I (IC50s = 2 and 98 nM, respectively); inhibits prenylation of FTase and GGTase I substrates HDJ2 and RAP1A in PSN-1 pancreatic tumor cells (EC50s = 92 and 6,760 nM, respectively); inhibits prenylation of the oncogenic protein KI-RAS in PSN-1 cells in a concentration-dependent manner; reduces HDJ2 and RAP1A prenylation ex vivo in dog PBMCs when administered at a dose of 35-50 mg/kg per day; has no effect on KI-RAS prenylation in patient-derived PBMCs; inhibits lectin-induced expression of the T cell activation markers CD71 and CD25 on human PMBCs (IC50s = 6.48 and 84.1 μM, respectively); inhibits IL-2-induced proliferation of CTLL-2 cells (IC50 = 0.81 μM)
Catalog No. 50-299-4563 Supplier Cayman Chemical Supplier No. 2294025 MG
A dual inhibitor of FTase and GGTase I (IC50s = 2 and 98 nM, respectively); inhibits prenylation of FTase and GGTase I substrates HDJ2 and RAP1A in PSN-1 pancreatic tumor cells (EC50s = 92 and 6,760 nM, respectively); inhibits prenylation of the oncogenic protein KI-RAS in PSN-1 cells in a concentration-dependent manner; reduces HDJ2 and RAP1A prenylation ex vivo in dog PBMCs when administered at a dose of 35-50 mg/kg per day; has no effect on KI-RAS prenylation in patient-derived PBMCs; inhibits lectin-induced expression of the T cell activation markers CD71 and CD25 on human PMBCs (IC50s = 6.48 and 84.1 μM, respectively); inhibits IL-2-induced proliferation of CTLL-2 cells (IC50 = 0.81 μM)