Learn More
TARGETMOL CHEMICALS INC L-778123 HYDROCHLORIDE 25MG
SDP
Catalog No. 502580348
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. The L-778123 hydrochloride is an inhibitor of FPTase (IC50 2 nM) and GGPTase-I (IC50 98 nM) in enzyme inhibition determination. purity: 98%
Catalog No. 50-258-0348 Supplier TARGETMOL CHEMICALS INC Supplier No. T318225MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. The L-778123 hydrochloride is an inhibitor of FPTase (IC50 2 nM) and GGPTase-I (IC50 98 nM) in enzyme inhibition determination. purity: 98%