Learn More
Medchemexpress LLC HY-12787 5mg Medchemexpress, L-779450 CAS:303727-31-3 Purity:>98%
SDP
Catalog No. 502081141
Encompass_Preferred
Medchemexpress, HY-12787 5mg L-779450 CAS:303727-31-3 L-779450 is a potent and selective B-Raf kinase inhibitor with a Kd of 2.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1141 Supplier Medchemexpress LLC Supplier No. HY127875MG
Medchemexpress, HY-12787 5mg L-779450 CAS:303727-31-3 L-779450 is a potent and selective B-Raf kinase inhibitor with a Kd of 2.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.