Learn More
TARGETMOL CHEMICALS INC L-796 778 1MG
SDP
Catalog No. 502877823
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. L-796,778 is an SST3 agonist and a selective ligand of SST3, which can increase the level of cAMP in the body.
Catalog No. 50-287-7823 Supplier TARGETMOL CHEMICALS INC Supplier No. T325111MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. L-796,778 is an SST3 agonist and a selective ligand of SST3, which can increase the level of cAMP in the body.