Learn More
Medchemexpress LLC L-796778 acetate | 2922280-21-3 | 1MG
SDP
Catalog No. 5000239314
Encompass_Preferred
L-796778 acetate is a selective agonist of the sst3 receptor In CHO-K1 cells expressing the hsst3 receptor L-796778 acetate is a partial agonist that inhibits Forskolin (HY-15371)-stimulated cAMP production with an IC50 value of 18 nM L-796778 acetate has anticonvulsant effect[1]
Catalog No. 50-002-39314 Supplier Medchemexpress LLC Supplier No. HY123335A1MG
L-796778 acetate is a selective agonist of the sst3 receptor In CHO-K1 cells expressing the hsst3 receptor L-796778 acetate is a partial agonist that inhibits Forskolin (HY-15371)-stimulated cAMP production with an IC50 value of 18 nM L-796778 acetate has anticonvulsant effect[1]