Learn More
Medchemexpress LLC L-796778 acetate | 2922280-21-3 | 98.0% | 644.72 | C31H44N6O9 | 5MG
SDP
Catalog No. 5000235483
Encompass_Preferred
L-796778 acetate is a selective agonist of the somatostatin receptor 3 (sst3) used for research applications. It functions as a partial agonist at the human sst3 receptor and inhibits forskolin-stimulated cAMP production in CHO-K1 cells with an IC50 of 18 nM; anticonvulsant activity has been reported. The compound is supplied as a solid and is intended for laboratory research use.
Catalog No. 50-002-35483 Supplier Medchemexpress LLC Supplier No. HY123335A5MG
L-796778 acetate is a selective agonist of the somatostatin receptor 3 (sst3) used for research applications. It functions as a partial agonist at the human sst3 receptor and inhibits forskolin-stimulated cAMP production in CHO-K1 cells with an IC50 of 18 nM; anticonvulsant activity has been reported. The compound is supplied as a solid and is intended for laboratory research use.