Learn More
Medchemexpress LLC L-803,087 trifluoroacetate | 217480-26-7 | MFCD09037334 | 99.7% | 485.53 g/mol | C25H29F2N5O3 | 5 MG
SDP
Catalog No. 5000264304
Encompass_Preferred
L-803087 is a potent, selective somatostatin sst4 receptor agonist used in receptor pharmacology and preclinical research. It demonstrates subnanomolar affinity and high selectivity, and is supplied as a stable solid suitable for in vitro formulation and testing.
  • Potent somatostatin sst4 receptor agonist (Ki = 0.7 nM) with >280-fold selectivity versus other somatostatin receptors.
  • High purity (99.7%) suitable for research applications.
  • White to light yellow solid; molecular weight 485.53 g/mol.
  • Soluble in DMSO (250 mg/mL); ultrasonic assistance recommended.
  • Stable as powder at -20°C for up to 3 years; in solvent: -80°C for 6 months or -20°C for 1 month.
  • Available in milligram packages, including 5 mg, and as a 10 mM solution in DMSO for reconstitution.
Catalog No. 50-002-64304 Supplier Medchemexpress LLC Supplier No. HY1084975MG
L-803087 is a potent, selective somatostatin sst4 receptor agonist used in receptor pharmacology and preclinical research. It demonstrates subnanomolar affinity and high selectivity, and is supplied as a stable solid suitable for in vitro formulation and testing.
  • Potent somatostatin sst4 receptor agonist (Ki = 0.7 nM) with >280-fold selectivity versus other somatostatin receptors.
  • High purity (99.7%) suitable for research applications.
  • White to light yellow solid; molecular weight 485.53 g/mol.
  • Soluble in DMSO (250 mg/mL); ultrasonic assistance recommended.
  • Stable as powder at -20°C for up to 3 years; in solvent: -80°C for 6 months or -20°C for 1 month.
  • Available in milligram packages, including 5 mg, and as a 10 mM solution in DMSO for reconstitution.