Learn More
Medchemexpress LLC L-803087 | 217480-26-7 | 99.7% | 485.53 | 1 MG
SDP
Catalog No. 5000346926
Encompass_Preferred
L-803087 is a potent and selective somatostatin sst4 receptor agonist with a Ki of 0.7 nM. It is >280-fold more selective than other somatostatin receptors. This compound facilitates AMPA-mediated hippocampal synaptic responses in vitro and increases kainate-induced seizures in mice.
  • Potent and selective somatostatin sst4 receptor agonist.
  • Greater than 280-fold higher selectivity for sst4 receptor compared to other somatostatin receptors.
  • Facilitates AMPA-mediated hippocampal synaptic responses in vitro.
  • Increases kainate-induced seizures in mice.
  • Does not inhibit secretion of growth hormone, insulin, or glucagon.
Catalog No. 50-003-46926 Supplier Medchemexpress LLC Supplier No. HY10849750MG
L-803087 is a potent and selective somatostatin sst4 receptor agonist with a Ki of 0.7 nM. It is >280-fold more selective than other somatostatin receptors. This compound facilitates AMPA-mediated hippocampal synaptic responses in vitro and increases kainate-induced seizures in mice.
  • Potent and selective somatostatin sst4 receptor agonist.
  • Greater than 280-fold higher selectivity for sst4 receptor compared to other somatostatin receptors.
  • Facilitates AMPA-mediated hippocampal synaptic responses in vitro.
  • Increases kainate-induced seizures in mice.
  • Does not inhibit secretion of growth hormone, insulin, or glucagon.