Learn More
Medchemexpress LLC L-803087 | 217480-26-7 | 99.7% | 485.53 | 10 MG
SDP
Catalog No. 5000346883
Encompass_Preferred
L-803087 is a potent and selective somatostatin sst4 receptor agonist with a Ki of 0.7 nM, showing >280-fold higher selectivity than other somatostatin receptors. It facilitates AMPA-mediated hippocampal synaptic responses in vitro and increases kainate-induced seizures in mice.
  • Potent and selective somatostatin sst4 receptor agonist
  • Facilitates AMPA-mediated hippocampal synaptic responses in vitro
  • Increases kainate-induced seizures in mice
Catalog No. 50-003-46883 Supplier Medchemexpress LLC Supplier No. HY10849710MG
L-803087 is a potent and selective somatostatin sst4 receptor agonist with a Ki of 0.7 nM, showing >280-fold higher selectivity than other somatostatin receptors. It facilitates AMPA-mediated hippocampal synaptic responses in vitro and increases kainate-induced seizures in mice.
  • Potent and selective somatostatin sst4 receptor agonist
  • Facilitates AMPA-mediated hippocampal synaptic responses in vitro
  • Increases kainate-induced seizures in mice