Learn More
Medchemexpress LLC L-826266 | 244101-03-9 | 98.0% | 570.88 | 50 MG
SDP
Catalog No. 5000384933
Encompass_Preferred
L-826266 is a selective and competitive EP3 receptor antagonist that can be used for convulsive disorders research. It also prevents PGE2-induced decrease of Na+,K+-ATPase activity in adult rat hippocampal slices and delays and increases the latency for clonic and generalized tonic-clonic seizures induced by PTZ in animal models.
  • Selective and competitive EP3 receptor antagonist
  • Can be used for convulsive disorders research
  • Prevents PGE2-induced decrease of Na+,K+-ATPase activity in adult rat hippocampal slices
  • Delays and increases the latency for clonic and generalized tonic-clonic seizures induced by PTZ in animal models
Catalog No. 50-003-84933 Supplier Medchemexpress LLC Supplier No. HY1936150MG
L-826266 is a selective and competitive EP3 receptor antagonist that can be used for convulsive disorders research. It also prevents PGE2-induced decrease of Na+,K+-ATPase activity in adult rat hippocampal slices and delays and increases the latency for clonic and generalized tonic-clonic seizures induced by PTZ in animal models.
  • Selective and competitive EP3 receptor antagonist
  • Can be used for convulsive disorders research
  • Prevents PGE2-induced decrease of Na+,K+-ATPase activity in adult rat hippocampal slices
  • Delays and increases the latency for clonic and generalized tonic-clonic seizures induced by PTZ in animal models