Learn More
Medchemexpress LLC HY-111271 10mg Medchemexpress, L 888607 CAS:860033-06-3 Purity:>98%
SDP
Catalog No. 501968831
Encompass_Preferred
Medchemexpress, HY-111271 10mg L 888607 CAS:860033-06-3 L 888607 is a potent, and selective CRTH2 (also known as DP2) agonist with a Ki of 0.8 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8831 Supplier Medchemexpress LLC Supplier No. HY11127110MG
Medchemexpress, HY-111271 10mg L 888607 CAS:860033-06-3 L 888607 is a potent, and selective CRTH2 (also known as DP2) agonist with a Ki of 0.8 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.