Learn More
Medchemexpress LLC L 888607 | 860033-06-3 | 99.87% | 375.84 | 10 MG
SDP
Catalog No. 5000344186
Encompass_Preferred
L 888607 is a potent, selective, stable, and orally active CRTH2 agonist. It has a high affinity for the human CRTH2 receptor with a Ki value of 4 nM. This product is suitable for research into various physiological events and metabolites.
  • Potent CRTH2 agonist.
  • Selective for CRTH2 receptor.
  • Stable.
  • Orally active.
  • High affinity for human CRTH2 receptor (Ki value of 4 nM).
  • Suitable for research of several physiological events and metabolites.
Catalog No. 50-003-44186 Supplier Medchemexpress LLC Supplier No. HY11127110MM/1ML
L 888607 is a potent, selective, stable, and orally active CRTH2 agonist. It has a high affinity for the human CRTH2 receptor with a Ki value of 4 nM. This product is suitable for research into various physiological events and metabolites.
  • Potent CRTH2 agonist.
  • Selective for CRTH2 receptor.
  • Stable.
  • Orally active.
  • High affinity for human CRTH2 receptor (Ki value of 4 nM).
  • Suitable for research of several physiological events and metabolites.