Learn More
Cayman Chemical L-a-AmInoadIpIc AcId 1g
SDP
Catalog No. 502968375
Encompass_Preferred
A gliotoxic analog of the excitatory neurotransmitter glutamate that competitively inhibits glutamine synthetase (Ki = 209 µM); actively accumulates in glial cells and inhibits the glutamate transporter with a (Ki = 192 µM)
Catalog No. 50-296-8375 Supplier Cayman Chemical Supplier No. 175791 G
A gliotoxic analog of the excitatory neurotransmitter glutamate that competitively inhibits glutamine synthetase (Ki = 209 µM); actively accumulates in glial cells and inhibits the glutamate transporter with a (Ki = 192 µM)